Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001433.3 | NP_001424.3 | 8 | 868 | ||
XM_017024347.1 | XP_016879836.1 | 8 | 2222 | |||
XM_017024348.1 | XP_016879837.1 | 7 | 2120 | |||
GenBank mRNA | AB209869.1 | - | 8 | 851 | ||
AF059198.1 | - | 8 | 851 | |||
AK292403.1 | - | 8 | 894 | |||
BC130405.1 | - | 8 | 801 | 40146436 | ||
BC130407.1 | - | 8 | 801 | 40146438 |