Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002000.3 | NP_001991.1 | 5 | 933 | ||
NM_133269.3 | NP_579803.1 | 5 | 867 | |||
NM_133271.3 | NP_579805.1 | 4 | 645 | |||
NM_133272.3 | NP_579806.1 | 4 | 897 | |||
NM_133273.3 | NP_579807.1 | 3 | 609 | |||
NM_133274.3 | NP_579808.1 | 4 | 823 | |||
NM_133277.3 | NP_579811.1 | 3 | 606 | |||
NM_133278.3 | NP_579812.1 | 4 | 831 | |||
XM_011526625.2 | XP_011524927.1 | 5 | 857 | |||
XM_017026473.1 | XP_016881962.1 | 5 | 906 | |||
XM_017026474.1 | XP_016881963.1 | 4 | 689 | |||
GenBank mRNA | AK290922.1 | - | 5 | 791 | ||
BC027953.1 | - | 5 | 760 | 5211453 | ||
D87853.1 | - | 5 | 670 | |||
D87854.1 | - | 4 | 448 | |||
D87855.1 | - | 4 | 700 | |||
D87856.1 | - | 3 | 412 | |||
D87857.1 | - | 4 | 626 | |||
D87858.1 | - | 5 | 526 | |||
D87859.1 | - | 6 | 748 | |||
D87860.1 | - | 4 | 490 | |||
D87861.1 | - | 3 | 409 | |||
D87862.1 | - | 4 | 634 | |||
DQ075334.1 | - | 5 | 736 | |||
DQ075335.1 | - | 5 | 736 | |||
DQ075336.1 | - | 5 | 736 | |||
DQ075337.1 | - | 5 | 736 | |||
DQ075338.1 | - | 5 | 736 | |||
DQ075339.1 | - | 5 | 736 | |||
S82919.1 | - | 4 | 448 | |||
U43677.1 | - | 4 | 453 | |||
U43774.1 | - | 5 | 675 | |||
X90753.1 | - | 2 | 197 | |||
X92558.1 | - | 4 | 700 | |||
X92559.1 | - | 4 | 634 | |||
X92560.1 | - | 5 | 670 | |||
X92561.1 | - | 4 | 448 | |||
X92562.1 | - | 3 | 409 |