Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000410.3 | NP_000401.1 | 2 | 288 | ||
NM_001300749.1 | NP_001287678.1 | 2 | 288 | |||
NM_139003.2 | NP_620572.1 | 2 | 288 | |||
NM_139004.2 | NP_620573.1 | 2 | 288 | |||
NM_139006.2 | NP_620575.1 | 2 | 288 | |||
XM_011514543.2 | XP_011512845.1 | 2 | 250 | |||
GenBank mRNA | AF079407.1 | - | 2 | 164 | ||
AF115264.1 | - | 2 | 134 | |||
AF115265.1 | - | 2 | 139 | |||
AF144242.1 | - | 2 | 251 | |||
AF149804.1 | - | 2 | 172 | |||
AJ249337.1 | - | 2 | 288 | |||
AK293393.1 | - | 2 | 140 | |||
AK300933.1 | - | 2 | 132 | |||
AK316502.1 | - | 2 | 140 | |||
BC074721.2 | - | 2 | 177 | 30915171 | ||
BC117201.1 | - | 2 | 163 | 40125752 | ||
BC117203.1 | - | 2 | 163 | 40125754 | ||
JN375754.1 | - | 2 | 89 | |||
U60319.1 | - | 2 | 349 |