Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KALRN SIRNA Assays ›
Gene Symbol:
KALRN
Gene Name:
kalirin, RhoGEF kinase
Gene Aliases:
ARHGEF24, CHD5, CHDS5, DUET, DUO, HAPIP, TRAD
Chromosome Location:
Chr.3: 124033341 - 124726325 on Build GRCh38
UniGene ID:
Hs.13281
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s200469
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KALRN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001024660.4
NP_001019831.2
10
1812
NM_001322988.1
NP_001309917.1
10
1812
NM_001322989.1
NP_001309918.1
10
1812
NM_001322990.1
NP_001309919.1
10
1812
NM_001322991.1
NP_001309920.1
10
1812
NM_001322992.1
NP_001309921.1
10
1812
NM_003947.5
NP_003938.1
10
1812
XM_006713810.3
XP_006713873.1
10
2057
XM_006713811.3
XP_006713874.1
10
2057
XM_006713812.3
XP_006713875.1
10
2057
XM_006713813.3
XP_006713876.1
10
2057
XM_006713814.3
XP_006713877.1
10
2057
XM_006713815.3
XP_006713878.1
10
2060
XM_011513279.2
XP_011511581.1
10
2057
XM_011513280.2
XP_011511582.1
10
2058
XM_011513281.2
XP_011511583.1
10
24706
XM_011513283.2
XP_011511585.1
10
2054
XM_011513285.2
XP_011511587.1
10
2053
XM_017007429.1
XP_016862918.1
10
2057
XM_017007430.1
XP_016862919.1
10
2057
XM_017007431.1
XP_016862920.1
10
2053
XM_017007432.1
XP_016862921.1
10
2053
XM_017007433.1
XP_016862922.1
10
2060
XM_017007434.1
XP_016862923.1
10
2060
XM_017007435.1
XP_016862924.1
10
2060
GenBank mRNA
AK314126.1
-
10
1812
U94190.1
-
10
1755
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
signal transduction
nervous system development
vesicle-mediated transport
regulation of Rho protein signal transduction
intracellular signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
ephrin receptor signaling pathway
regulation of small GTPase mediated signal transduction
protein serine/threonine kinase activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0