Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015488.4 | NP_056303.3 | 7 | 846 | ||
NM_022572.4 | NP_072094.1 | 6 | 719 | |||
XM_017003771.1 | XP_016859260.1 | 7 | 753 | |||
XM_017003772.1 | XP_016859261.1 | 6 | 744 | |||
GenBank mRNA | AB033010.1 | - | 7 | 687 | ||
AK289867.1 | - | 7 | 754 | |||
AK309308.1 | - | 549 | ||||
AL137675.1 | - | 5 | 1345 | |||
AY358753.1 | - | 6 | 639 | |||
BC002937.2 | - | 6 | 630 | 3956630 | ||
BC036457.2 | - | 7 | 748 | 4797574 | ||
BI764334.1 | - | 6 | 705 |