Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244808.1 | NP_001231737.1 | 7 | 865 | ||
NM_001244810.1 | NP_001231739.1 | 7 | 762 | |||
NM_001244812.1 | NP_001231741.1 | 4 | 390 | |||
NM_001244814.1 | NP_001231743.1 | 3 | 559 | |||
NM_001244815.1 | NP_001231744.1 | 2 | 500 | |||
NM_001244816.1 | NP_001231745.1 | 6 | 667 | |||
NM_032682.5 | NP_116071.2 | 7 | 762 | |||
XM_005264735.3 | XP_005264792.1 | 2 | 451 | |||
XM_005264736.3 | XP_005264793.1 | 2 | 500 | |||
XM_005264737.4 | XP_005264794.1 | 7 | 771 | |||
XM_006713102.2 | XP_006713165.1 | 6 | 723 | |||
XM_006713103.2 | XP_006713166.1 | 4 | 460 | |||
XM_006713104.2 | XP_006713167.1 | 5 | 693 | |||
XM_011533584.2 | XP_011531886.1 | 7 | 833 | |||
XM_011533585.2 | XP_011531887.1 | 6 | 685 | |||
XM_011533588.2 | XP_011531890.1 | 2 | 283 | |||
XM_017006165.1 | XP_016861654.1 | 6 | 604 | |||
XM_017006166.1 | XP_016861655.1 | 6 | 700 | |||
XM_017006167.1 | XP_016861656.1 | 2 | 500 | |||
GenBank mRNA | AF146696.1 | - | 5 | 499 | ||
AF250920.1 | - | 6 | 667 | |||
AK027264.1 | - | 2 | 132 | |||
AK074321.1 | - | 3 | 558 | |||
AK092383.1 | - | 4 | 390 | |||
AK122710.1 | - | 7 | 865 | |||
BC131720.1 | - | 4 | 387 | |||
BX647682.1 | - | 2 | 148 | |||
DA634017.1 | - | 2 | 500 |