Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023946.3 | NP_076435.1 | 3 | 334 | ||
NM_177457.4 | NP_803252.1 | 3 | 334 | |||
NM_177476.3 | NP_803429.1 | 3 | 353 | |||
NM_177477.3 | NP_803430.1 | 3 | 467 | |||
GenBank mRNA | AF321824.1 | - | 3 | 293 | ||
BC014560.2 | - | 2 | 520 | |||
BG701653.1 | - | 3 | 352 | |||
BI603715.1 | - | 3 | 471 | |||
BM172253.1 | - | 3 | 352 | |||
BQ685632.1 | - | 3 | 282 | |||
DB501423.1 | - | 3 | 468 | |||
HY110957.1 | - | 3 | 351 |