Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKBIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201612.2 | NP_963906.1 | 3 | 1129 | ||
NM_201613.2 | NP_963907.1 | 2 | 1011 | |||
GenBank mRNA | AJ539426.1 | - | 3 | 889 | ||
AJ539427.1 | - | 2 | 771 | |||
AK123069.1 | - | 2 | 915 | |||
BC029415.1 | - | 2 | 771 | |||
BC051372.1 | - | 2 | 740 | |||
BC107572.1 | - | 2 | 532 |