Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308353.1 | NP_001295282.1 | 4 | 804 | ||
NM_005855.3 | NP_005846.1 | 3 | 743 | |||
XM_017003152.1 | XP_016858641.1 | 3 | 665 | |||
XM_017003153.1 | XP_016858642.1 | 3 | 973 | |||
XM_017003154.1 | XP_016858643.1 | 3 | 750 | |||
XM_017003156.1 | XP_016858645.1 | 3 | 691 | |||
GenBank mRNA | AJ001014.1 | - | 3 | 643 | ||
BC000548.2 | - | 3 | 635 | 3163522 |