Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHAMMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003521.1 | 8 | 1616 | |||
GenBank mRNA | BC018097.1 | - | 6 | 805 | ||
BC044610.1 | - | 6 | 939 | |||
BC048987.1 | - | 8 | 1676 | |||
DQ309039.1 | - | 6 | 795 | |||
DQ309040.1 | - | 8 | 1616 |
Gene Symbol: WHAMMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036650.1 | 5 | 1109 | |||
GenBank mRNA | BI910970.1 | - | 3 | 379 | ||
BX643549.1 | - | 2 | 166 |
Gene Symbol: WHAMMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026589.1 | 7 | 1510 | |||
GenBank mRNA | AK091254.1 | - | 2 | 192 | ||
BC066982.1 | - | 8 | 1356 | |||
BC150524.1 | - | 6 | 866 |