Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005858.3 | NP_005849.1 | 10 | 1269 | ||
XM_011527624.2 | XP_011525926.1 | 10 | 1362 | |||
XM_017026141.1 | XP_016881630.1 | 10 | 1414 | |||
GenBank mRNA | AK303640.1 | - | 7 | 896 | ||
AK308251.1 | - | 9 | 1163 | |||
AK309529.1 | - | 7 | 797 | |||
AL050160.1 | - | 4 | 197 | |||
BC037270.2 | - | 10 | 1259 | 4837309 | ||
Y11997.1 | - | 10 | 1260 |