Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005862.2 | NP_005853.2 | 22 | 2517 | ||
XM_011512329.2 | XP_011510631.1 | 24 | 2721 | |||
XM_011512331.2 | XP_011510633.1 | 23 | 2651 | |||
XM_017005523.1 | XP_016861012.1 | 21 | 2395 | |||
XM_017005524.1 | XP_016861013.1 | 22 | 2591 | |||
XM_017005525.1 | XP_016861014.1 | 10 | 1089 | |||
GenBank mRNA | AB210023.1 | - | 22 | 2482 | ||
AK304381.1 | - | 11 | 1057 | |||
BC064699.1 | - | 22 | 2492 | 30342305 | ||
CR749247.1 | - | 25 | 2891 | |||
Z75330.1 | - | 22 | 2625 |