Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005862.2 | NP_005853.2 | 11 | 1365 | ||
XM_011512329.2 | XP_011510631.1 | 13 | 1569 | |||
XM_011512331.2 | XP_011510633.1 | 12 | 1499 | |||
XM_017005523.1 | XP_016861012.1 | 10 | 1243 | |||
XM_017005524.1 | XP_016861013.1 | 11 | 1439 | |||
GenBank mRNA | AB210023.1 | - | 11 | 1330 | ||
AK304381.1 | - | 1 | 13 | |||
AK309934.1 | - | 12 | 1465 | |||
BC064699.1 | - | 11 | 1340 | 30342305 | ||
CR749247.1 | - | 14 | 1739 | |||
Z75330.1 | - | 11 | 1473 |