Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014981.1 | NP_001014981.1 | 9, 10 | 1204 | ||
GenBank mRNA | AF188123.1 | - | 8, 9 | 966 | ||
AF188124.1 | - | 8, 9 | 966 | |||
AK052730.1 | - | 9, 10 | 1205 | |||
AK122304.1 | - | 1, 2 | 93 | |||
AK154451.1 | - | 9, 10 | 1184 | |||
BC026618.1 | - | 9, 10 | 1123 | |||
BC125573.1 | - | 9, 10 | 1072 | 40130088 | ||
BC137785.1 | - | 9, 10 | 1072 | 8860804 |