Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001796.4 | NP_001001796.1 | 3 | 283 | ||
GenBank mRNA | AK039633.1 | - | 3 | 284 | ||
AY116506.1 | - | 2 | 152 | |||
BC132613.1 | - | 3 | 252 | 40130890 | ||
BC145917.1 | - | 3 | 252 | 40131220 | ||
BC171926.1 | - | 3 | 252 | 9053913 | ||
EU670677.1 | - | 4 | 403 | |||
EU670678.1 | - | 4 | 403 |