Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080639.3 | NP_542370.3 | 3 | 407 | ||
GenBank mRNA | AF282730.1 | - | 3 | 338 | ||
AK038820.1 | - | 3 | 406 | |||
AK040814.1 | - | 3 | 406 | |||
AK079347.1 | - | 3 | 408 | |||
AK140367.1 | - | 3 | 407 | |||
BC064046.1 | - | 3 | 387 | 875422 |