Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Egln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053208.4 | NP_444438.2 | 2 | 951 | ||
GenBank mRNA | AF340231.1 | - | 1 | 693 | ||
AF453879.1 | - | 2 | 873 | |||
AJ310547.1 | - | 2 | 922 | |||
AK075582.1 | - | 2 | 903 | |||
AK152056.1 | - | 2 | 912 | |||
AK152223.1 | - | 2 | 909 | |||
AK159206.1 | - | 2 | 904 | |||
AK166661.1 | - | 2 | 932 | |||
AK167477.1 | - | 2 | 904 | |||
BC023299.1 | - | 2 | 885 | 5135579 | ||
BC051436.1 | - | 1 | 669 | 6312638 | ||
BC086764.1 | - | 2 | 890 | 5685311 |