Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLVRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000713.2 | NP_000704.1 | 1, 2 | 207 | ||
GenBank mRNA | AK312137.1 | - | 1, 2 | 134 | ||
BC109371.1 | - | 1, 2 | 124 | 5229026 | ||
BG340491.1 | - | 1, 2 | 207 | |||
D26308.1 | - | 1, 2 | 155 | |||
D32143.1 | - | 1, 2 | 186 | |||
EU794593.1 | - | 1, 2 | 207 |