Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007666.3 | NP_031692.2 | 12 | 2217 | ||
GenBank mRNA | BC107008.1 | - | 11 | 1958 | 40047565 | |
BC107013.1 | - | 11 | 1958 | 40048308 | ||
D82029.1 | - | 11 | 1958 | |||
U67399.1 | - | 4 | 525 |