Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021273.4 | NP_067248.1 | 2 | 272 | ||
GenBank mRNA | AK002467.1 | - | 2 | 235 | ||
AK014299.1 | - | 2 | 215 | |||
AK148885.1 | - | 2 | 232 | |||
AK152388.1 | - | 2 | 232 | |||
AK153484.1 | - | 2 | 232 | |||
AK161990.1 | - | 2 | 232 | |||
AK165779.1 | - | 1 | 492 | |||
AK166980.1 | - | 2 | 235 | |||
AK167034.1 | - | 2 | 235 | |||
BC015271.1 | - | 2 | 202 | 4225384 | ||
BC058257.1 | - | 1 | 472 | |||
BC106109.1 | - | 2 | 199 | 6395794 |