Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcnka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146307.1 | NP_001139779.1 | 4 | 538 | ||
NM_024412.3 | NP_077723.3 | 5 | 542 | |||
GenBank mRNA | AF124848.1 | - | 4 | 635 | ||
AY373832.1 | - | 4 | 493 | |||
BC037077.1 | - | 5 | 546 | 5042736 |