Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007720.2 | NP_031746.1 | 2 | 923 | ||
GenBank mRNA | BC101966.1 | - | 1 | 867 | 40044931 | |
BC101967.1 | - | 1 | 867 | 40044932 | ||
BC103566.1 | - | 1 | 867 | 40044933 | ||
BC103567.1 | - | 1 | 867 | 40044935 |