Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dffb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007859.4 | NP_031885.3 | 2 | 243 | ||
GenBank mRNA | AB009377.1 | - | 2 | 151 | ||
AK036094.1 | - | 2 | 220 | |||
AK077619.1 | - | 2 | 289 | |||
AK089267.1 | - | 2 | 220 | |||
BC053052.1 | - | 2 | 173 | 6403143 |