Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252188.1 | NP_001239117.1 | 9 | 1761 | ||
NM_001252189.1 | NP_001239118.1 | 7 | 1517 | |||
NM_010148.3 | NP_034278.1 | 8 | 1553 | |||
GenBank mRNA | AF057286.1 | - | 6 | 1008 | ||
AK036331.1 | - | 7 | 1491 | |||
AK139234.1 | - | 9 | 1749 | |||
AK139793.1 | - | 9 | 1684 | |||
AK160782.1 | - | 7 | 1518 | |||
AK173093.1 | - | 9 | 1737 | |||
BC039138.1 | - | 8 | 1517 | 2647379 |