Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166368.1 | NP_001159840.1 | 3 | 289 | ||
NM_008020.3 | NP_032046.1 | 3 | 304 | |||
GenBank mRNA | AK144284.1 | - | 3 | 305 | ||
AK165394.1 | - | 3 | 269 | |||
BC031824.1 | - | 3 | 258 | 4196363 | ||
BC053692.1 | - | 3 | 294 | 6395841 | ||
CF949699.1 | - | 2 | 108 | |||
DV655383.1 | - | 3 | 289 |