Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glul | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008131.4 | NP_032157.2 | 7 | 1222 | ||
GenBank mRNA | AK137969.1 | - | 7 | 1209 | ||
AK159106.1 | - | 7 | 1208 | |||
AK160670.1 | - | 7 | 1209 | |||
AK162685.1 | - | 7 | 1207 | |||
AK168493.1 | - | 7 | 1206 | |||
AY044241.1 | - | 7 | 1206 | |||
BC015086.1 | - | 7 | 1206 | 4224865 | ||
U09114.1 | - | 6 | 1098 | |||
X16314.1 | - | 7 | 1183 |