Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HSPA8 SIRNA Assays ›
Gene Symbol:
Hspa8
Gene Name:
heat shock protein 8
Gene Aliases:
2410008N15Rik, Hsc70, Hsc71, Hsc73, Hsp73, Hspa10
Chromosome Location:
Chr.9: 40801273 - 40805199 on Build GRCm38
UniGene ID:
Mm.290774
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s201481
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hspa8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031165.4
NP_112442.2
5
729
GenBank mRNA
AK004608.1
-
4
1018
AK035286.1
-
5
727
AK075935.1
-
5
726
AK081701.1
-
5
728
AK098074.1
-
5
726
AK145416.1
-
5
649
AK145579.1
-
5
723
AK146547.1
-
5
730
AK146708.1
-
5
724
AK146985.1
-
5
727
AK147864.1
-
5
727
AK149954.1
-
5
724
AK150474.1
-
5
721
AK150498.1
-
5
721
AK150701.1
-
5
731
AK150958.1
-
5
724
AK151041.1
-
5
727
AK151065.1
-
5
727
AK151127.1
-
5
730
AK151287.1
-
5
727
AK151435.1
-
5
727
AK151516.1
-
5
733
AK151537.1
-
5
724
AK151741.1
-
5
727
AK151775.1
-
5
729
AK151808.1
-
5
730
AK151865.1
-
5
727
AK151892.1
-
5
729
AK151948.1
-
5
730
AK151997.1
-
5
727
AK152315.1
-
5
730
AK152598.1
-
5
727
AK152697.1
-
5
727
AK152703.1
-
5
727
AK152768.1
-
5
730
AK152803.1
-
5
730
AK153032.1
-
5
729
AK153834.1
-
5
727
AK157831.1
-
5
727
AK159479.1
-
5
727
AK162775.1
-
5
727
AK163987.1
-
5
727
AK164000.1
-
5
727
AK166545.1
-
5
729
AK166721.1
-
5
724
AK166767.1
-
5
727
AK166776.1
-
5
730
AK166808.1
-
5
727
AK166830.1
-
5
725
AK166846.1
-
5
727
AK166861.1
-
5
730
AK166908.1
-
5
730
AK166910.1
-
5
727
AK166913.1
-
5
727
AK166933.1
-
5
727
AK167121.1
-
5
730
AK167122.1
-
5
727
AK167134.1
-
5
725
AK167163.1
-
5
729
AK167218.1
-
5
727
AK167229.1
-
5
728
AK167414.1
-
5
727
AK167845.1
-
5
730
AK167910.1
-
5
730
AK168435.1
-
5
730
AK168492.1
-
5
729
AK168511.1
-
3
265
AK168519.1
-
5
727
AK168542.1
-
5
729
AK168577.1
-
5
727
AK168625.1
-
5
727
AK168654.1
-
5
727
AK168711.1
-
5
727
AK168750.1
-
5
730
AK168776.1
-
5
730
AK168887.1
-
5
729
AK168934.1
-
5
730
AK169093.1
-
5
727
AK169179.1
-
5
727
AK169236.1
-
5
727
AK169293.1
-
5
741
AK169329.1
-
5
730
AK169594.1
-
5
726
AK171434.1
-
5
728
AK178143.1
-
2
208
AK184919.1
-
2
208
AK190509.1
-
2
208
AK192182.1
-
2
209
AK192272.1
-
2
208
AK193850.1
-
2
208
AK193882.1
-
2
208
AK198438.1
-
2
208
AK198532.1
-
3
269
AK199142.1
-
2
208
AK199354.1
-
2
208
AK199410.1
-
2
241
AK199504.1
-
2
209
AK200094.1
-
2
208
AK200099.1
-
2
209
AK200309.1
-
2
208
AK200579.1
-
2
208
AK202859.1
-
2
208
AK203911.1
-
2
208
AK204890.1
-
2
208
AK205054.1
-
2
208
AK205187.1
-
2
208
AK206134.1
-
2
209
AK209289.1
-
2
208
AK210012.1
-
3
274
AK211519.1
-
2
208
AK212081.1
-
3
272
AK212914.1
-
2
228
AK216159.1
-
2
208
AK216732.1
-
2
208
AK216875.1
-
2
209
BC006722.1
-
5
751
3711399
BC066191.1
-
5
683
30467052
BC085486.1
-
5
715
30540914
BC089322.1
-
5
708
30538640
BC089457.1
-
5
686
30628505
BC094900.1
-
5
643
30063483
BC106169.1
-
3
407
BC106193.1
-
5
711
6329283
M19141.1
-
5
713
U27129.1
-
5
678
X05837.1
-
1
81
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
protein folding
RNA splicing
protein refolding
negative regulation of transcription, DNA-templated
ATP metabolic process
positive regulation of mRNA splicing, via spliceosome
chaperone mediated protein folding requiring cofactor
regulation of cell cycle
chaperone-mediated protein folding
clathrin coat disassembly
negative regulation of fibril organization
nucleotide binding
G-protein coupled receptor binding
receptor binding
protein binding
ATP binding
ATPase activity
enzyme binding
MHC class II protein complex binding
heat shock protein binding
ubiquitin protein ligase binding
ATPase activity, coupled
ADP binding
poly(A) RNA binding
unfolded protein binding
C3HC4-type RING finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0