Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005180.8 | NP_005171.4 | 9 | 1128 | ||
GenBank mRNA | BC011652.2 | - | 9 | 1127 | 4138748 | |
BT019450.1 | - | 8 | 622 | |||
CR456911.1 | - | 8 | 622 | |||
L13689.1 | - | 9 | 1101 |
Gene Symbol: COMMD3-BMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204062.1 | NP_001190991.1 | 13 | 1086 | ||
GenBank mRNA | AK313235.1 | - | 13 | 1061 |