Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206871.1 | NP_996754.1 | 1 | 297 |
Gene Symbol: Ifna13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177347.2 | NP_796321.1 | 1 | 371 | ||
GenBank mRNA | AY190047.1 | - | 1 | 371 | ||
BC120724.1 | - | 1 | 327 | 40129647 | ||
BC120750.1 | - | 1 | 327 | 40129673 |
Gene Symbol: Ifna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010505.2 | NP_034635.2 | 1 | 297 | ||
GenBank mRNA | BC120910.1 | - | 1 | 297 | 40091152 | |
BC120911.1 | - | 1 | 297 | 40091153 |