Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il18bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010531.1 | NP_034661.1 | 7 | 792 | ||
GenBank mRNA | AF110802.1 | - | 4 | 2226 | ||
AF110803.1 | - | 6 | 719 | |||
AF122907.1 | - | 7 | 792 | |||
AK003370.1 | - | 7 | 808 | |||
AK008452.1 | - | 6 | 723 | |||
AK009721.1 | - | 6 | 719 | |||
BC018332.1 | - | 6 | 702 | 4007411 |