Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008459.2 | NP_032485.2 | 1 | 70 | ||
GenBank mRNA | AF110492.1 | - | 1 | 70 |
Gene Symbol: Klra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289604.1 | NP_001276533.1 | 2 | 238 | ||
NM_001289605.1 | NP_001276534.1 | 2 | 272 | |||
NM_010648.3 | NP_034778.2 | 2 | 238 | |||
GenBank mRNA | AK159595.1 | - | 2 | 187 | ||
U56404.1 | - | 2 | 113 |