Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCIRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006019.3 | NP_006010.2 | 12 | 1488 | ||
NM_006053.3 | NP_006044.1 | 7 | 1247 | |||
XM_005273709.3 | XP_005273766.1 | 13 | 1647 | |||
XM_011544726.2 | XP_011543028.1 | 12 | 1387 | |||
XM_017017089.1 | XP_016872578.1 | 12 | 1483 | |||
XM_017017090.1 | XP_016872579.1 | 11 | 1826 | |||
GenBank mRNA | AF025374.1 | - | 7 | 1247 | ||
BC018133.1 | - | 12 | 1467 | 3905446 | ||
BC032465.1 | - | 12 | 1460 | 5210733 | ||
U45285.1 | - | 12 | 1416 |