Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133225.3 | NP_573488.2 | 1, 2 | 329 | ||
GenBank mRNA | AF022770.3 | - | 1, 2 | 330 | ||
AK030371.1 | - | 1, 2 | 330 | |||
AK167272.1 | - | 1, 2 | 311 | |||
AK168799.1 | - | 1, 2 | 301 | |||
BC050121.1 | - | 1, 2 | 300 | |||
BC060602.1 | - | 1, 2 | 284 | 5706182 |