Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LANCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136574.1 | NP_001130046.1 | 8, 9 | 1118 | ||
NM_001136575.1 | NP_001130047.1 | 8, 9 | 1113 | |||
NM_006055.2 | NP_006046.1 | 8, 9 | 1207 | |||
XM_005246243.2 | XP_005246300.1 | 8, 9 | 1228 | |||
GenBank mRNA | AK309295.1 | - | 5, 6 | 410 | ||
AK312381.1 | - | 8, 9 | 1096 | |||
BC028685.2 | - | 8, 9 | 1118 | 4829574 | ||
CR541777.1 | - | 7, 8 | 1043 | |||
Y11395.1 | - | 8, 9 | 1147 |