Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcpt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010782.3 | NP_034912.3 | 5 | 1957 | ||
GenBank mRNA | AF007120.1 | - | 4 | 621 | ||
AY007568.1 | - | 5 | 1957 |
Gene Symbol: Mcpt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M55617.1 | - | 766 |