Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013603.2 | NP_038631.1 | 2 | 245 | ||
GenBank mRNA | AK140209.1 | - | 2 | 246 | ||
BC059725.1 | - | 2 | 143 | 6600430 |