Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138660.2 | NP_619601.2 | 12 | 2033 | ||
GenBank mRNA | AF526276.1 | - | 12 | 2007 | ||
AJ292072.1 | - | 2244 | ||||
AK037399.1 | - | 6 | 628 | |||
AK139608.1 | - | 12 | 2072 | |||
AK145886.1 | - | 12 | 1807 | |||
BC034533.1 | - | 9 | 1677 | |||
BC060672.1 | - | 12 | 1858 | |||
BC141296.1 | - | 12 | 2146 | 9055908 |