Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011293.2 | NP_035423.2 | 1, 2 | 117 | ||
GenBank mRNA | AK158610.1 | - | 1, 2 | 118 | ||
AK158644.1 | - | 1, 2 | 107 | |||
BC042939.1 | - | 1, 2 | 74 | 6476299 | ||
D85818.1 | - | 1, 2 | 71 | |||
DV065741.1 | - | 151 |