Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sncg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011430.3 | NP_035560.1 | 3 | 283 | ||
GenBank mRNA | AA690669.1 | - | 3 | 397 | ||
AF017255.1 | - | 3 | 262 | |||
AJ786826.1 | - | 3 | 196 | |||
AK141509.1 | - | 3 | 267 | |||
BC028508.1 | - | 3 | 274 | 1448798 |