Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172136.1 | NP_001165607.1 | 5 | 601 | ||
NM_172456.3 | NP_766044.1 | 4 | 566 | |||
GenBank mRNA | AK028887.1 | - | 4 | 530 | ||
AK034654.1 | - | 5 | 563 | |||
AK051300.1 | - | 5 | 565 | |||
AK148426.1 | - | 4 | 547 | |||
AK162566.1 | - | 4 | 529 | |||
AK163555.1 | - | 4 | 530 | |||
BC052171.1 | - | 3 | 372 | |||
BC059920.1 | - | 5 | 544 | |||
BC064730.1 | - | 5 | 616 | |||
BC094589.1 | - | 2 | 3052 | |||
BC119024.1 | - | 4 | 530 | 8733773 | ||
BC144729.1 | - | 5 | 565 |