Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166480.1 | NP_001159952.1 | 3 | 424 | ||
NM_009279.3 | NP_033305.1 | 3 | 306 | |||
GenBank mRNA | AK003044.1 | - | 3 | 286 | ||
AK007921.1 | - | 3 | 262 | |||
AK013723.1 | - | 3 | 268 | |||
AK142600.1 | - | 1 | 2706 | |||
AK160089.1 | - | 3 | 272 | |||
BC008172.1 | - | 3 | 227 | 3481948 | ||
BC012971.1 | - | 2 | 1807 | |||
BC019419.1 | - | 2 | 1757 | |||
BI989805.1 | - | 3 | 424 | |||
BQ747160.1 | - | 3 | 209 | |||
CJ144362.1 | - | 3 | 311 | |||
X90582.1 | - | 3 | 387 |