Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172509.3 | NP_766097.1 | 4 | 1827 | ||
GenBank mRNA | AK029942.1 | - | 4 | 1803 | ||
AK034583.1 | - | 4 | 1803 | |||
AK083583.1 | - | 4 | 1828 | |||
AK140606.1 | - | 4 | 1802 | |||
AK162568.1 | - | 4 | 1856 | |||
BC139070.1 | - | 4 | 1729 | 8862092 | ||
BC141201.1 | - | 4 | 1729 | 9055813 |