Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp10c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167578.1 | NP_001161050.1 | 12 | 1593 | ||
GenBank mRNA | AK076614.1 | - | 12 | 1539 | ||
AK132653.1 | - | 13 | 1689 | |||
BC061173.1 | - | 12 | 1564 | 6772123 |
Gene Symbol: Tcp10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011553.4 | NP_035683.2 | 12 | 1493 | ||
GenBank mRNA | AK161455.1 | - | 12 | 1464 | ||
BC050771.1 | - | 12 | 1459 | 6773681 | ||
BC052353.1 | - | 12 | 1608 | 6773668 | ||
X58169.1 | - | 12 | 1359 | |||
X58171.1 | - | 12 | 1466 |
Gene Symbol: Tcp10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009341.2 | NP_033367.2 | 12 | 1499 | ||
GenBank mRNA | AK077165.1 | - | 12 | 1508 | ||
BC141264.1 | - | 11 | 1370 | 9055876 | ||
M22602.1 | - | 1 | 32 | |||
X58170.1 | - | 11 | 2125 |