Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009341.2 | NP_033367.2 | 8 | 1018 | ||
GenBank mRNA | AK077165.1 | - | 8 | 1027 | ||
BC141264.1 | - | 7 | 889 | 9055876 | ||
M22599.1 | - | 7 | 1010 | |||
M22601.1 | - | 7 | 922 | |||
X58170.1 | - | 7 | 1644 |
Gene Symbol: Tcp10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011553.4 | NP_035683.2 | 8 | 1012 | ||
GenBank mRNA | AK161455.1 | - | 8 | 983 | ||
BC050771.1 | - | 8 | 978 | 6773681 | ||
BC052353.1 | - | 8 | 1127 | 6773668 | ||
M22600.1 | - | 7 | 860 | |||
X58169.1 | - | 8 | 878 | |||
X58171.1 | - | 8 | 985 |