Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285988.1 | NP_001272917.1 | 17 | 2468 | ||
NM_001285989.1 | NP_001272918.1 | 17 | 2450 | |||
NM_001285990.1 | NP_001272919.1 | 16 | 2373 | |||
NM_145436.2 | NP_663411.2 | 17 | 2453 | |||
GenBank mRNA | AK148278.1 | - | 17 | 2309 | ||
AK161143.1 | - | 6 | 829 | |||
AK161471.1 | - | 17 | 2318 | |||
BC157955.1 | - | 17 | 2467 | 9088128 | ||
BC172100.1 | - | 17 | 2467 | 9054386 | ||
BC172149.1 | - | 16 | 2372 |