Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081170.1 | NP_001074639.1 | 22 | 2469 | ||
NM_001291444.1 | NP_001278373.1 | 22 | 2475 | |||
NM_001291445.1 | NP_001278374.1 | 21 | 2379 | |||
GenBank mRNA | AK030003.1 | - | 17 | 1838 | ||
AK036581.1 | - | 21 | 2355 | |||
AK054000.1 | - | 22 | 2450 | |||
AK122326.1 | - | 20 | 2158 | |||
AK193976.1 | - | 1 | 196 | |||
BC043302.1 | - | 3 | 386 | |||
BC059076.1 | - | 7 | 1039 |