Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpbpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009411.4 | NP_033437.2 | 4 | 354 | ||
GenBank mRNA | AK005406.1 | - | 4 | 354 | ||
AK005417.1 | - | 4 | 355 | |||
AK005438.1 | - | 4 | 354 | |||
AK005446.1 | - | 4 | 355 | |||
AK005454.1 | - | 4 | 356 | |||
AK005499.1 | - | 4 | 355 | |||
AK011401.1 | - | 4 | 355 | |||
AK014443.1 | - | 4 | 356 | |||
AK018797.1 | - | 4 | 355 | |||
AK018800.1 | - | 4 | 354 | |||
AK018802.1 | - | 4 | 354 | |||
AK018803.1 | - | 4 | 355 | |||
AK018806.1 | - | 4 | 355 | |||
AK018807.1 | - | 4 | 355 | |||
AK018811.1 | - | 4 | 355 | |||
AK018812.1 | - | 4 | 355 | |||
AK018813.1 | - | 4 | 355 | |||
AK018814.1 | - | 4 | 355 | |||
AK018815.1 | - | 4 | 354 | |||
AK018819.1 | - | 4 | 355 | |||
AK018821.1 | - | 4 | 355 | |||
AK018822.1 | - | 4 | 356 | |||
AK018823.1 | - | 4 | 355 | |||
AK018825.1 | - | 4 | 356 | |||
AK018826.1 | - | 4 | 355 | |||
AK028071.1 | - | 4 | 354 | |||
AK160140.1 | - | 4 | 348 | |||
AY034575.1 | - | 4 | 330 | |||
BC099391.1 | - | 4 | 341 | 30787380 | ||
X17071.1 | - | 4 | 346 |