Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aim1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162970.1 | NP_001156442.1 | 16 | 4093 | ||
GenBank mRNA | AK155034.1 | - | 4 | 444 | ||
AK165037.1 | - | 3 | 368 | |||
BC042709.1 | - | 11 | 1114 | |||
BC150738.1 | - | 15 | 4022 | 9087649 |