Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313892.1 | NP_001300821.1 | 4, 5 | 645 | ||
NM_001313893.1 | NP_001300822.1 | 4, 5 | 1405 | |||
NM_002128.5 | NP_002119.1 | 4, 5 | 648 | |||
GenBank mRNA | AK057120.1 | - | 4, 5 | 511 | ||
AK122825.1 | - | 4, 5 | 1407 | |||
AK291494.1 | - | 4, 5 | 619 | |||
AX795278.1 | - | 4, 5 | 540 | |||
AY377859.1 | - | 3, 4 | 464 | |||
BC003378.1 | - | 4, 5 | 540 | 2901382 | ||
BC030981.1 | - | 4, 5 | 559 | 4041682 | ||
BC066889.1 | - | 4, 5 | 543 | 5268698 | ||
BC067732.1 | - | 4, 5 | 563 | 4818524 | ||
BC141844.1 | - | 4, 5 | 541 | 8860319 | ||
BT006940.1 | - | 3, 4 | 464 | |||
BT020159.1 | - | 3, 4 | 464 | |||
CR456863.1 | - | 3, 4 | 464 | |||
CR749614.1 | - | 4, 5 | 636 | |||
D63874.1 | - | 4, 5 | 540 | |||
X12597.1 | - | 4, 5 | 516 |
Gene Symbol: CTCFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC137482.1 | - | 1 | 499 | 9021482 | |
BC137483.1 | - | 1 | 499 | 9021483 |