Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177693.3 | NP_808361.1 | 4 | 412 | ||
GenBank mRNA | AK029354.1 | - | 4 | 413 | ||
AK145594.1 | - | 1521 | ||||
AK164958.1 | - | 4 | 411 | |||
BC119253.1 | - | 4 | 393 | 8734002 | ||
BC119255.1 | - | 4 | 393 | 8734004 |